Isopropyltriphenylphosphoniumiodid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

377481-25G
SIAL377481-25GEA 106 EUR
SIAL377481-25G
Isopropyltriphenylphosphoniumiodid, Sigma-Aldrich®
Isopropyltriphenylphosphoniumiodid
Formel: C₂₁H₂₂IP
Molecular Weight: 432,29 g/mol
MDL: MFCD00031595
CAS-Nummer: 24470-78-8
EINECS: 246-281-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR