(1-Heptyl)triphenylphosphoniumbromid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Heptyltriphenylphosphonium bromide
M-Clarity™ Qualitätsstufe = MQ100
377538-50G
SIAL377538-50GEA
109
EUR
SIAL377538-50G
(1-Heptyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(1-Heptyl)triphenylphosphoniumbromid
Formel:
H₃C(CH₂)₆P(C₆H₅)₃Br Molecular Weight: 441.39 g/mol Schmelzpunkt: 176…177 °C |
MDL:
MFCD00050249 CAS-Nummer: 13423-48-8 EINECS: 236-539-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...