3-Brom-1-(trimethylsilyl)-1-propin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane
M-Clarity™ Qualitätsstufe = MQ100
377775-10G
377775-1G
SIAL377775-10GEA
259
EUR
SIAL377775-10G
SIAL377775-1G
3-Brom-1-(trimethylsilyl)-1-propin, Sigma-Aldrich®
3-Brom-1-(trimethylsilyl)-1-propin
Formel:
C₆H₁₁BrSi Molecular Weight: 191.14 g/mol |
MDL:
MFCD00134460 CAS-Nummer: 38002-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...