4,4'-Diaminobenzophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(4-aminophenyl)methanone, 4,4′-Diaminobenzophenone

M-Clarity™ Qualitätsstufe = MQ100

378259-5G
SIAL378259-5GEA 112 EUR
SIAL378259-5G
4,4'-Diaminobenzophenon, Sigma-Aldrich®
4,4'-Diaminobenzophenon
Formel: C₁₃H₁₂N₂O
Molecular Weight: 212,25 g/mol
MDL: MFCD00038138
CAS-Nummer: 611-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR