3,3-Diethoxypropiononitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,3-Diethoxypropionitrile, 3,3-Diethoxypropanenitrile, Cyanoacetaldehyde diethyl acetal

M-Clarity™ Qualitätsstufe = MQ100

378615-25ML
SIAL378615-25MLEA 130 EUR
SIAL378615-25ML
3,3-Diethoxypropiononitril, Sigma-Aldrich®
3,3-Diethoxypropiononitril
Formel: C₇H₁₃NO₂
Molecular Weight: 143,19 g/mol
MDL: MFCD00040558
CAS-Nummer: 2032-34-0
EINECS: 217-988-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR