Dinatriumtetrachloropalladat ≥99,995% (Metall-Basis), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Natriumtetrachloropalladat(II), Natrium-Palladium(II)-chlorid

M-Clarity™ Qualitätsstufe = MQ100

379808-1G
SIAL379808-1GEA 117 EUR
SIAL379808-1G
Dinatriumtetrachloropalladat ≥99,995% (Metall-Basis), Sigma-Aldrich®
Dinatriumtetrachloropalladat
Formel: Cl₄Na₂Pd
Molecular Weight: 294,21 g/mol
MDL: MFCD00003487
CAS-Nummer: 13820-53-6
EINECS: 237-502-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR