Gold(III)chlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Goldtrichlorid
M-Clarity™ Qualitätsstufe = MQ100
379948-1G
379948-250MG
SIAL379948-1GEA
342
EUR
SIAL379948-1G
SIAL379948-250MG
Gold(III)chlorid, Sigma-Aldrich®
Gold(III)chlorid
Formel:
AuCl₃ Molecular Weight: 303.32 g/mol Schmelzpunkt: 287…288 °C Dichte: 4.3 g/cm³ (20 °C) |
MDL:
MFCD00014172 CAS-Nummer: 13453-07-1 EINECS: 236-623-1 UN: 3260 ADR: 8 III Merck Index: 13,04536 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...