Cobalt(II)carbonat Hydrat, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

379956-5G
SIAL379956-5GEA 137 EUR
SIAL379956-5G
Cobalt(II)carbonat Hydrat, Sigma-Aldrich®
Cobalt(II)carbonat Hydrat
Formel: CCoO₃·H₂O
MDL: MFCD00149654
CAS-Nummer: 57454-67-8
EINECS: 208-169-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR