Chlordiisopropyloctylsilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

380695-25ML
SIAL380695-25MLEA 398 EUR
SIAL380695-25ML
Chlordiisopropyloctylsilan, Sigma-Aldrich®
Chlordiisopropyloctylsilan
Formel: C₁₄H₃₁ClSi
MDL: MFCD00134528
CAS-Nummer: 117559-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR