Bis(cyclopentadienyl)chrom, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chromocene, Bis(Cyclopentadienyl)Chromium(Ii)

M-Clarity™ Qualitätsstufe = MQ100

381004-1G
SIAL381004-1GEA 141 EUR
SIAL381004-1G
Bis(cyclopentadienyl)chrom, Sigma-Aldrich®
Bis(cyclopentadienyl)chrom
Formel: C₁₀H₁₀Cr
Molecular Weight: 182,19 g/mol
MDL: MFCD00013750
CAS-Nummer: 1271-24-5
EINECS: 215-036-4
UN: 3391
ADR: 4.2,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR