2-Aminoterephthalsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Aminoterephthalic Acid

M-Clarity™ Qualitätsstufe = MQ100

381071-25G
SIAL381071-25GEA 93.2 EUR
SIAL381071-25G
2-Aminoterephthalsäure, Sigma-Aldrich®
2-Aminoterephthalsäure
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
MDL: MFCD00134536
CAS-Nummer: 10312-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR