3,6-Difluorphthalsäureanhydrid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,6-Difluor-1,2-benzoldicarbonsäureanhydrid

M-Clarity™ Qualitätsstufe = MQ200

381128-500MG
SIAL381128-500MGEA 100 EUR
SIAL381128-500MG
3,6-Difluorphthalsäureanhydrid, Sigma-Aldrich®
3,6-Difluorphthalsäureanhydrid
Formel: C₈H₂F₂O₃
MDL: MFCD00134537
CAS-Nummer: 652-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR