6-Fluor-m-toluonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

381330-5G
SIAL381330-5GEA 106 EUR
SIAL381330-5G
6-Fluor-m-toluonitril, Sigma-Aldrich®
6-Fluor-m-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135,14 g/mol
MDL: MFCD00134542
CAS-Nummer: 64113-84-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR