N-Cyclohexylmaleimid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

381543-5G
SIAL381543-5GEA 167 EUR
SIAL381543-5G
N-Cyclohexylmaleimid, Sigma-Aldrich®
N-Cyclohexylmaleimid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,22 g/mol
MDL: MFCD00043904
CAS-Nummer: 1631-25-0
EINECS: 216-630-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR