3,3'-Dimethoxybenzil, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Bis(3-methoxyphenyl)ethandion

M-Clarity™ Qualitätsstufe = MQ100

381802-5G
SIAL381802-5GEA 110 EUR
SIAL381802-5G
3,3'-Dimethoxybenzil, Sigma-Aldrich®
3,3'-Dimethoxybenzil
Formel: C₁₆H₁₄O₄
Molecular Weight: 270.28 g/mol
MDL: MFCD00038221
CAS-Nummer: 40101-17-5
EINECS: 254-793-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR