2-Brom-5-nitrobenzoesäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
381845-5G
SIAL381845-5GEA
36.4
EUR
SIAL381845-5G
2-Brom-5-nitrobenzoesäure, Sigma-Aldrich®
2-Bromo-5-nitrobenzoic acid
Formel:
C₇H₄BrNO₄ Molecular Weight: 246,02 g/mol Siedepunkt: 371 °C (1013 hPa) Schmelzpunkt: 178…180 °C |
MDL:
MFCD00134558 CAS-Nummer: 943-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...