2-(Hydroxymethyl)-12-krone-4, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

382655-1G
SIAL382655-1GEA 131 EUR
SIAL382655-1G
2-(Hydroxymethyl)-12-krone-4, Sigma-Aldrich®
2-(Hydroxymethyl)-12-krone-4
Formel: C₉H₁₈O₅
MDL: MFCD00067524
CAS-Nummer: 75507-26-5
EINECS: 278-224-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR