1,3-Difluor-2-nitrobenzol, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 2,6-Difluoronitrobenzene

M-Clarity™ Qualitätsstufe = MQ200

382957-5G
SIAL382957-5GEA 48.4 EUR
SIAL382957-5G
1,3-Difluor-2-nitrobenzol, Sigma-Aldrich®
1,3-Difluor-2-nitrobenzol
Formel: C₆H₃F₂NO₂
Molecular Weight: 159.09 g/mol
MDL: MFCD00192035
CAS-Nummer: 19064-24-5
EINECS: 242-793-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR