Tantal(V)methanolat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tantal(V)methoxid

M-Clarity™ Qualitätsstufe = MQ100

383244-5G
SIAL383244-5GEA 197 EUR
SIAL383244-5G
Tantal(V)methanolat, Sigma-Aldrich®
Tantal(V)methanolat
Formel: C₅H₁₅O₅Ta
MDL: MFCD00058817
CAS-Nummer: 865-35-0
UN: 3181
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR