Tantal(V)butanolat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

383333-10G
SIAL383333-10GEA 512 EUR
SIAL383333-10G
Tantal(V)butanolat, Sigma-Aldrich®
Tantal(V)butanolat
Formel: C₂₀H₄₅O₅Ta
MDL: MFCD00078035
CAS-Nummer: 51094-78-1
EINECS: 256-962-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR