5'-Brom-2'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(5-Brom-2-hydroxyphenyl)ethan-1-on, 2-Acetyl-4-bromphenol

M-Clarity™ Qualitätsstufe = MQ100

383406-10G
SIAL383406-10GEA 64.3 EUR
SIAL383406-10G
5'-Brom-2'-hydroxyacetophenon, Sigma-Aldrich®
5'-Brom-2'-hydroxyacetophenon
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
MDL: MFCD00191850
CAS-Nummer: 1450-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR