3-Formyl-6-methylchromon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Methyl-4-oxo-4H-chromene-3-carbaldehyde

M-Clarity™ Qualitätsstufe = MQ100

383473-5G
SIAL383473-5GEA 60.8 EUR
SIAL383473-5G
3-Formyl-6-methylchromon, Sigma-Aldrich®
3-Formyl-6-methylchromon
Formel: C₁₁H₈O₃
Molecular Weight: 188,1794 g/mol
MDL: MFCD00138943
CAS-Nummer: 42059-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR