3-Cyanchromon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chromone-3-carbonitrile, 4-Oxo-4H-chromene-3-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

383481-5G
SIAL383481-5GEA 79 EUR
SIAL383481-5G
3-Cyanchromon, Sigma-Aldrich®
3-Cyanchromon
Formel: C₁₀H₅NO₂
Molecular Weight: 171,16 g/mol
MDL: MFCD00052604
CAS-Nummer: 50743-17-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR