Tri(2-furyl)phosphin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tri(furan-2-yl)phosphine, Tri-2-furylphosphine

M-Clarity™ Qualitätsstufe = MQ100

383767-1G
SIAL383767-1GEA 95.7 EUR
SIAL383767-1G
Tri(2-furyl)phosphin, Sigma-Aldrich®
Tri(2-furyl)phosphine
Formel: C₁₂H₉O₃P
Molecular Weight: 232,17 g/mol
MDL: MFCD00151857
CAS-Nummer: 5518-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR