2-Brom-5-methoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

384003-5G
SIAL384003-5GEA 23.5 EUR
SIAL384003-5G
2-Brom-5-methoxybenzoesäure, Sigma-Aldrich®
2-Bromo-5-methoxybenzoic acid
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
MDL: MFCD00020214
CAS-Nummer: 22921-68-2
EINECS: 245-329-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR