2,3,5,6-Tetrafluor-4-(trifluormethyl)phenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxy-2,3,5,6-tetrafluorobenzotrifluoride

M-Clarity™ Qualitätsstufe = MQ100

384070-1G
SIAL384070-1GEA 69 EUR
SIAL384070-1G
2,3,5,6-Tetrafluor-4-(trifluormethyl)phenol, Sigma-Aldrich®
2,3,5,6-Tetrafluor-4-(trifluormethyl)phenol
Formel: C₇HF₇O
MDL: MFCD00155984
CAS-Nummer: 2787-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR