2,3,5-Trichlorpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,5-TCP

M-Clarity™ Qualitätsstufe = MQ200

384275-50G
SIAL384275-50GEA 86.8 EUR
SIAL384275-50G
2,3,5-Trichlorpyridin, Sigma-Aldrich®
2,3,5-Trichlorpyridin
Formel: C₅H₂Cl₃N
Molecular Weight: 182,44 g/mol
MDL: MFCD00043007
CAS-Nummer: 16063-70-0
EINECS: 407-270-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR