8-Bromchinolin, Sigma-Aldrich®

Lieferant: Merck
Achtung

384348-500MG
SIAL384348-500MGEA 95 EUR
SIAL384348-500MG
8-Bromchinolin, Sigma-Aldrich®
8-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00191859
CAS-Nummer: 16567-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR