Chlortrimethylsilan 1 mol/l in THF, Sigma-Aldrich®

Lieferant: Merck

Synonyme: TMCS, Trimethylsilylchlorid, Trimethylchlorsilan, TMSC

M-Clarity™ Qualitätsstufe = MQ100

384410-100ML
SIAL384410-100MLEA 51 EUR
SIAL384410-100ML
Chlortrimethylsilan 1 mol/l in THF, Sigma-Aldrich®
Chlortrimethylsilan
Formel: C₃H₉ClSi
Molecular Weight: 108,64 g/mol
MDL: MFCD00000502
CAS-Nummer: 75-77-4
EINECS: 200-900-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR