1,4-Difluor-2,5-dimethoxybenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

385352-5G
SIAL385352-5GEA 127 EUR
SIAL385352-5G
1,4-Difluor-2,5-dimethoxybenzol, Sigma-Aldrich®
1,4-Difluoro-2,5-dimethoxybenzene
Formel: C₈H₈F₂O₂
MDL: MFCD00142553
CAS-Nummer: 199866-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR