4-Brom-3-methylanisol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-5-methoxytoluol, 1-Brom-4-methoxy-2-methylbenzol

M-Clarity™ Qualitätsstufe = MQ100

385409-25ML
SIAL385409-25MLEA 125 EUR
SIAL385409-25ML
4-Brom-3-methylanisol, Sigma-Aldrich®
4-Brom-3-methylanisol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00060687
CAS-Nummer: 27060-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR