3,6-Dioxaoctamethylendiamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

385506-100ML 385506-500ML
SIAL385506-100MLEA 86.2 EUR
SIAL385506-100ML SIAL385506-500ML
3,6-Dioxaoctamethylendiamin, Sigma-Aldrich®
3,6-Dioxaoctamethylendiamin
Formel: C₆H₁₆N₂O₂
MDL: MFCD00040474
CAS-Nummer: 929-59-9
EINECS: 213-203-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR