Methyl-3,5-dinitrobenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dinitrobenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

386367-25G
SIAL386367-25GEA 100 EUR
SIAL386367-25G
Methyl-3,5-dinitrobenzoat, Sigma-Aldrich®
Methyl-3,5-dinitrobenzoat
Formel: C₈H₆N₂O₆
Molecular Weight: 226,15 g/mol
MDL: MFCD00017016
CAS-Nummer: 2702-58-1
EINECS: 220-289-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR