4,4'-(Hexafluorisopropyliden)diphthalsäureanhydrid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

386448-25G 386448-5G
SIAL386448-25GEA 354 EUR
SIAL386448-25G SIAL386448-5G
4,4'-(Hexafluorisopropyliden)diphthalsäureanhydrid, Sigma-Aldrich®
4,4'-(Hexafluorisopropyliden)diphthalsäureanhydrid
Formel: C₈H₄O₃
MDL: MFCD00039143
CAS-Nummer: 1107-00-2
EINECS: 201-607-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR