(4-Hydroxyphenyl)acetamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxyphenylacetamide

M-Clarity™ Qualitätsstufe = MQ100

387738-5G
SIAL387738-5GEA 16.4 EUR
SIAL387738-5G
(4-Hydroxyphenyl)acetamid, Sigma-Aldrich®
(4-Hydroxyphenyl)acetamid
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00017145
CAS-Nummer: 17194-82-0
EINECS: 241-235-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR