5-Brompentylacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Bromamylacetat, Essigsäure(5-bromamyl)ester, Essigsäure(5-brompentyl)ester
M-Clarity™ Qualitätsstufe = MQ200
387843-25ML
SIAL387843-25MLEA
118
EUR
SIAL387843-25ML
5-Brompentylacetat, Sigma-Aldrich®
5-Brompentylacetat
Formel:
C₇H₁₃BrO₂ Molecular Weight: 209,08 g/mol |
MDL:
MFCD00039204 CAS-Nummer: 15848-22-3 EINECS: 239-961-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...