Dimethylamin 2 mol/l in Methanol, Sigma-Aldrich®

Lieferant: Merck

388246-100ML
SIAL388246-100MLEA 60.9 EUR
SIAL388246-100ML
Dimethylamin 2 mol/l in Methanol, Sigma-Aldrich®
Dimethylamin
Formel: C₂H₇N
Molecular Weight: 45,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008288
CAS-Nummer: 124-40-3
EINECS: 204-697-4
UN: 1992
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR