1-Acetyl-4-piperidon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Acetyl-4-piperidone

388254-5ML
SIAL388254-5MLEA 167 EUR
SIAL388254-5ML
1-Acetyl-4-piperidon, Sigma-Aldrich®
1-Acetyl-4-piperidon
Formel: C₇H₁₁NO₂
Molecular Weight: 141,17 g/mol
MDL: MFCD00006190
CAS-Nummer: 32161-06-1
EINECS: 250-937-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR