Tributylacetylcitrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tributyl O-Acetylcitrate, Acetyltributylcitrate, Acetyl tributyl citrate, Tributyl 2-acetoxypropane-1,2,3-tricarboxylate, O-Acetylcitric acid tributyl ester

MQ100

388378-500ML
SIAL388378-500MLEA 65.6 EUR
SIAL388378-500ML
Tributylacetylcitrat, Sigma-Aldrich®
Tributylacetylcitrat
Formel: C₂₀H₃₄O₈
Molecular Weight: 402.49 g/mol
MDL: MFCD00043554
CAS-Nummer: 77-90-7
EINECS: 201-067-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR