2-(Hydroxymethyl)-15-krone-5, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Hydroxymethyl)-15-crown-5-ether

M-Clarity™ Qualitätsstufe = MQ100

388424-1G
SIAL388424-1GEA 95.5 EUR
SIAL388424-1G
2-(Hydroxymethyl)-15-krone-5, Sigma-Aldrich®
2-(Hydroxymethyl)-15-krone-5
Formel: C₁₁H₂₂O₆
Molecular Weight: 250.29 g/mol
MDL: MFCD00192215
CAS-Nummer: 75507-25-4
EINECS: 278-222-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR