2-(Aminomethyl)-18-krone-6, Sigma-Aldrich®

Lieferant: Merck

388432-1G 388432-250MG
SIAL388432-1GEA 741 EUR
SIAL388432-1G SIAL388432-250MG
2-(Aminomethyl)-18-krone-6, Sigma-Aldrich®
2-(Aminomethyl)-18-krone-6
Formel: C₁₃H₂₇NO₆
CAS-Nummer: 83585-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR