Hexakis(brommethyl)benzol, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 1,2,3,4,5,6-Hexakis(bromomethyl)benzene

M-Clarity™ Qualitätsstufe = MQ100

388459-5G
SIAL388459-5GEA 140 EUR
SIAL388459-5G
Hexakis(brommethyl)benzol, Sigma-Aldrich®
Hexakis(brommethyl)benzol
Formel: C₁₂H₁₂Br₆
Molecular Weight: 635,65 g/mol
MDL: MFCD00182539
CAS-Nummer: 3095-73-6
EINECS: 221-443-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR