2,4,6-Tris(brommethyl)mesitylen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3,5-Tris(bromomethyl)-2,4,6-trimethylbenzene, 2,4,6-Tris(bromomethyl)mesitylene

M-Clarity™ Qualitätsstufe = MQ100

388467-5G
SIAL388467-5GEA 79.9 EUR
SIAL388467-5G
2,4,6-Tris(brommethyl)mesitylen, Sigma-Aldrich®
2,4,6-Tris(brommethyl)mesitylen
Formel: C₁₂H₁₅Br₃
Molecular Weight: 398,96 g/mol
MDL: MFCD00192554
CAS-Nummer: 21988-87-4
EINECS: 244-697-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR