5,8-Dihydroxy-1,4-naphthochinon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Naphthazarin, 5,8-Dihydroxynaphthalene-1,4-dione
M-Clarity™ Qualitätsstufe = MQ100
388513-1G
SIAL388513-1GEA
269
EUR
SIAL388513-1G
5,8-Dihydroxy-1,4-naphthochinon, Sigma-Aldrich®
5,8-Dihydroxy-1,4-naphthochinon
Formel:
C₁₀H₆O₄ Molecular Weight: 190,16 g/mol |
MDL:
MFCD00001685 CAS-Nummer: 475-38-7 EINECS: 207-495-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...