(3-Benzyloxypropyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: [3-(Benzyloxy)propyl]triphenylphosphonium Bromide

389196-5G
SIAL389196-5GEA 165 EUR
SIAL389196-5G
(3-Benzyloxypropyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(3-Benzyloxypropyl)triphenylphosphoniumbromid
Formel: C₂₈H₂₈BrOP
Molecular Weight: 491.41 g/mol
MDL: MFCD00191781
CAS-Nummer: 54314-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR