4'-Butylacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Butylphenyl)ethanon, 4-Butyl-1-acetylbenzol, p-Butylacetophenon

M-Clarity™ Qualitätsstufe = MQ100

389250-5ML
SIAL389250-5MLEA 60.7 EUR
SIAL389250-5ML
4'-Butylacetophenon, Sigma-Aldrich®
4'-Butylacetophenon
Formel: C₁₂H₁₆O
Molecular Weight: 176,26 g/mol
MDL: MFCD00017500
CAS-Nummer: 37920-25-5
EINECS: 253-715-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR