5-Amino-4-cyan-1-phenyl-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

389595-1G
SIAL389595-1GEA 33.2 EUR
SIAL389595-1G
5-Amino-4-cyan-1-phenyl-1H-pyrazol, Sigma-Aldrich®
5-Amino-4-cyano-1-phenyl-1H-pyrazole
Formel: C₁₀H₈N₄
Molecular Weight: 184,20 g/mol
MDL: MFCD00020730
CAS-Nummer: 5334-43-0
EINECS: 226-253-9
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR