Pentylisocyanat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Pentyl isocyanate
M-Clarity™ Qualitätsstufe = MQ100
389951-5G
SIAL389951-5GEA
92.4
EUR
SIAL389951-5G
Pentylisocyanat, Sigma-Aldrich®
Pentylisocyanat
Formel:
C₆H₁₁NO Molecular Weight: 113.16 g/mol Siedepunkt: 136…137 °C (1013 hPa) Dichte: 0.878 g/cm³ (20 °C) |
MDL:
MFCD00144846 CAS-Nummer: 3954-13-0 UN: 3080 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...