4-Acetamido-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Acetamido-TEMPO), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

390380-25G
SIAL390380-25GEA 1240 EUR
SIAL390380-25G
4-Acetamido-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Acetamido-TEMPO), Sigma-Aldrich®
4-Acetamido-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Acetamido-TEMPO)
Formel: C₁₁H₂₁N₂O₂
MDL: MFCD00043593
CAS-Nummer: 14691-89-5
EINECS: 423-840-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR