3,5-Bis(trifluormethyl)-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Bis(trifluormethyl)-1H-pyrazol

M-Clarity™ Qualitätsstufe = MQ200

390399-1G
SIAL390399-1GEA 38 EUR
SIAL390399-1G
3,5-Bis(trifluormethyl)-1H-pyrazol, Sigma-Aldrich®
3,5-Bis(trifluormethyl)-1H-pyrazol
Formel: C₅H₂F₆N₂
Molecular Weight: 204.07 g/mol
Siedepunkt: 148
MDL: MFCD00153672
CAS-Nummer: 14704-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR